Tisk…

Furfuryl mercaptan ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B22380.14 B22380.22
B22380.14EA 1600 CZK
B22380.14 B22380.22
Furfuryl mercaptan ≥98%
Furfuryl mercaptan

Purity: 98+% CAS No.: 98-02-2 Molecular Formula: C5H6OS Formula Weight: 114.17 Flash Point: 45°(113°F) Refractive Index: 1.5315 Odour: Strong Boiling Point: 155° Beilstein Reference: 383594 UN No.: 1228 MDL No.: MFCD00003254

Vzorec: C₅H₆OS
Bod varu: 155 °C
Hustota: 1,132
Bod vzplanutí: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003254
CAS číslo: 98-02-2
EINECS: 202-628-2
UN: 1228
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR