Tisk…

2-Methyl-1-butanethiol ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B21949.06 B21949.14
B21949.06EA 1760 CZK
B21949.06 B21949.14
2-Methyl-1-butanethiol ≥99%
2-Methyl-1-butanethiol

Purity: 99% CAS No.: 1878-18-8 Molecular Formula: C5H12S Formula Weight: 104.22 Flash Point: 19°(67°F) Refractive Index: 1.4480 Boiling Point: 116-117° UN No.: 1993 MDL No.: MFCD00004883

Vzorec: C₅H₁₂S
Bod varu: 116…117 °C
Hustota: 0,845
Bod vzplanutí: 19 °C (67 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004883
CAS číslo: 1878-18-8
EINECS: 217-515-3
UN: 1993
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR