Tisk…

Dimethyl sulphone ≥99%

Dodavatel: Thermo Fisher Scientific

B21747.0B B21747.30
B21747.0BEA 4950 CZK
B21747.0B B21747.30
Dimethyl sulphone ≥99%
Dimethyl sulphone

Purity: 99% CAS No.: 67-71-0 Molecular Formula: C2H6O2S Formula Weight: 94.13 Flash Point: 143°(289°F) Odour: Odourless Boiling Point: 238° Beilstein Reference: 1737717 Merck Reference: 14,3258 MDL No.: MFCD00007566

Vzorec: C₂H₆O₂S
Bod varu: 238 °C
Bod tání: 108…111 °C
Hustota: 1,45
Bod vzplanutí: 143 °C (289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007566
CAS číslo: 67-71-0
EINECS: 200-665-9
Merck Index: 14,3258

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR