2-Thiopheneethanol ≥98%
Dodavatel: Thermo Fisher Scientific
B21562.06
B21562.14
B21562.22
B21562.06EA
1320
CZK
B21562.06
B21562.14
B21562.22
2-Thiopheneethanol ≥98%
2-Thiopheneethanol
Purity: 98%
CAS No.: 5402-55-1
Molecular Formula: C6H8OS
Formula Weight: 128.19
Flash Point: 101°(213°F)
Refractive Index: 1.5510
Boiling Point: 107-108°/12mm
Beilstein Reference: 106985
MDL No.: MFCD00005462
Vzorec:
C₆H₈OS Bod varu: 107…108 °C (12 mmHg) Hustota: 1,150 Bod vzplanutí: 101 °C (213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005462 CAS číslo: 5402-55-1 EINECS: 226-452-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...