Tisk…

2,5-Dichloro-p-phenylenediamine 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2,5-Dichlorobenzene-1,4-diamine, 1,4-Diamino-2,5-dichlorobenzene, 2,5-Dichloro-1,4-phenylenediamine

B21494.14 B21494.06
B21494.14EA 4230 CZK
B21494.14 B21494.06
2,5-Dichloro-p-phenylenediamine 99%
2,5-Dichloro-p-phenylenediamine

Purity: 99% CAS No.: 20103-09-7 Molecular Formula: C6H6Cl2N2 Formula Weight: 177.03 Melting Point: 164-168° MDL No.: MFCD00007902

Vzorec: C₆H₆Cl₂N₂
MW: 177,03 g/mol
Bod tání: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00007902
CAS číslo: 20103-09-7
EINECS: 243-512-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR