Tisk…

Methylová žluť indicator

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Dimethyl yellow, Sudan Yellow, 4-(Dimethylamino)azobenzene, 4-(N,N-Dimethylamino)azobenzene

B21145.14 B21145.22 B21145.36
B21145.14EA 810 CZK
B21145.14 B21145.22 B21145.36
Methylová žluť indicator
Methylová žluť

CAS No.: 60-11-7 Molecular Formula: C14H15N3 Formula Weight: 225.30 Odour: Odourless Melting Point: 110-112° dec. Solubility: Insoluble in water. Soluble in alcohol. Beilstein Reference: 746016 Merck Reference: 14,3229 UN No.: 2811 MDL No.: MFCD00008308

Vzorec: C₁₄H₁₅N₃
MW: 225.29
Bod varu: 371
Bod tání: 110…112 °C (rozkl.)
Hustota: 1.2233 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008308
CAS číslo: 60-11-7
EINECS: 200-455-7
UN: 2811
ADR: 6.1 III
Merck Index: 14,3229

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR