Tisk…

5-Chloro-2-methylindole ≥98%

Dodavatel: Thermo Fisher Scientific

B21036.03 B21036.06
B21036.03EA 1240 CZK
B21036.03 B21036.06
5-Chloro-2-methylindole ≥98%
5-Chloro-2-methylindole

Purity: 98% CAS No.: 1075-35-0 Molecular Formula: C9H8ClN Formula Weight: 165.62 Melting Point: 110-114° MDL No.: MFCD00005619

Vzorec: C₉H₈ClN
MW: 165.62
Bod varu: 130…140 °C (0.1 mmHg)
Bod tání: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00005619
CAS číslo: 1075-35-0
EINECS: 214-052-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR