Tisk…

Propyl propionate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Propionic acid propyl ester

B20680.AK B20680.AU
B20680.AKEA 1900 CZK
B20680.AK B20680.AU
Propyl propionate ≥99%
Propyl propionate

Purity: 99% CAS No.: 106-36-5 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 19°(67°F) Refractive Index: 1.3930 Melting Point: -76° Boiling Point: 122-124° Merck Reference: 14,7867 UN No.: 3272 MDL No.: MFCD00009373

Vzorec: C₆H₁₂O₂
Bod varu: 122…124 °C
Bod tání: –76 °C
Hustota: 0,881
Bod vzplanutí: 19 °C (67 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009373
CAS číslo: 106-36-5
EINECS: 203-389-7
UN: 3272
ADR: 3,II
Merck Index: 14,7867

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR