α-Bromo-p-toluonitrile ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B20641.06
B20641.14
B20641.06EA
1610
CZK
B20641.06
B20641.14
α-Bromo-p-toluonitrile ≥98%
α-Bromo-p-toluonitrile
Purity: 98%
CAS No.: 17201-43-3
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Melting Point: 113-117°
Beilstein Reference: 636717
UN No.: 2923
MDL No.: MFCD00001829
Vzorec:
C₈H₆BrN Bod varu: 135…137 °C (0,6 mmHg) Bod tání: 113…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001829 CAS číslo: 17201-43-3 EINECS: 241-246-0 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...