2-Fluoro-3-methylbenzyl bromide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-(Bromomethyl)-2-fluoro-3-methylbenzene
B20538.03
B20538.06
B20538.03EA
1430
CZK
B20538.03
B20538.06
2-Fluoro-3-methylbenzyl bromide ≥97%
2-Fluoro-3-methylbenzyl bromide
Purity: 97%
CAS No.: 151412-12-3
Molecular Formula: C8H8BrF
Formula Weight: 203.06
Refractive Index: 1.5480
Boiling Point: 105-106°/10mm
UN No.: 3265
MDL No.: MFCD00042470
Vzorec:
C₈H₈BrF MW: 203.05 Bod varu: 105…106 °C (10 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00042470 CAS číslo: 151412-12-3 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...