N-Aminophthalimide 94%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Aminoisoindoline-1,3-dione
A19914.06
A19914.09
A19914.18
A19914.06EA
1610
CZK
A19914.06
A19914.09
A19914.18
N-Aminophthalimide 94%
N-Aminophthalimide
Purity: 94%
CAS No.: 1875-48-5
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 195° dec.
Beilstein Reference: 383756
MDL No.: MFCD00005895
Vzorec:
C₈H₆N₂O₂ MW: 162,15 g/mol Bod varu: 391 °C (1013 hPa) Bod tání: ∼ 195 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005895 CAS číslo: 1875-48-5 EINECS: 217-505-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...