9-Decen-1-ol ≥90%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1-Decen-10-ol
A19490.14
A19490.22
A19490.14EA
2440
CZK
A19490.14
A19490.22
9-Decen-1-ol ≥90%
9-Decen-1-ol
Purity: 90+%
CAS No.: 13019-22-2
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 104°(219°F)
Refractive Index: 1.4480
Odour: Pleasant
Melting Point: -13°
Boiling Point: 235°
Beilstein Reference: 1750928
MDL No.: MFCD00002992
Vzorec:
C₁₀H₂₀O Bod varu: 235 °C Bod tání: –13 °C Hustota: 0,847 Bod vzplanutí: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002992 CAS číslo: 13019-22-2 EINECS: 235-878-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...