Tisk…

Isopropyl chloroacetate ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

A19070.AC A19070.AP
A19070.ACEA 1130 CZK
A19070.AC A19070.AP
Isopropyl chloroacetate ≥99%
Isopropyl chloroacetate

Purity: 99% CAS No.: 105-48-6 Molecular Formula: C5H9ClO2 Formula Weight: 136.58 Refractive Index: 1.4205 Boiling Point: 149-150° UN No.: 2947 MDL No.: MFCD00040410

Vzorec: C₅H₉ClO₂
MW: 136,58 g/mol
Bod varu: 149…150 °C
Bod tání: –112 °C
Hustota: 1,08
Storage Temperature: Ambient
MDL Number: MFCD00040410
CAS číslo: 105-48-6
EINECS: 203-301-7
UN: 2947
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR