2,4,5-Trimethylthiazole ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A19046.06
A19046.14
A19046.06EA
1080
CZK
A19046.06
A19046.14
2,4,5-Trimethylthiazole ≥98%
2,4,5-Trimethylthiazole
Purity: 98%
CAS No.: 13623-11-5
Molecular Formula: C6H9NS
Formula Weight: 127.21
Flash Point: 64°(147°F)
Refractive Index: 1.5090
Boiling Point: 164-166°
Beilstein Reference: 107148
MDL No.: MFCD00005332
Vzorec:
C₆H₉NS Bod varu: 164…166 °C Hustota: 1,043 Bod vzplanutí: 64 °C (147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005332 CAS číslo: 13623-11-5 EINECS: 237-107-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...