5-Iodouracil ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Iodopyrimidine-2,4(1H,3H)-dione, Iodouracil
A18994.09
A18994.18
A18994.09EA
1770
CZK
A18994.09
A18994.18
5-Iodouracil ≥97%
5-Iodouracil
Purity: 97%
CAS No.: 696-07-1
Molecular Formula: C4H3IN2O2
Formula Weight: 237.99
Melting Point: 274-276°
Beilstein Reference: 4891
Vzorec:
C₄H₃IN₂O₂ MW: 237,98 g/mol Bod varu: 401 °C (1013 hPa) Bod tání: 274…276 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006020 CAS číslo: 696-07-1 EINECS: 211-788-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...