6-Fluoro-m-anisaldehyde ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Fluoro-5-methoxybenzaldehyde, 6-Fluoro-3-anisaldehyde
A18904.06
A18904.14
A18904.06EA
2480
CZK
A18904.06
A18904.14
6-Fluoro-m-anisaldehyde ≥97%
6-Fluoro-m-anisaldehyde
Purity: 97%
CAS No.: 105728-90-3
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Flash Point: 107°(224°F)
Refractive Index: 1.5330
Boiling Point: 80°/0.9mm
Beilstein Reference: 4379088
MDL No.: MFCD00070795
Vzorec:
C₈H₇FO₂ MW: 154,14 g/mol Bod varu: 80 °C (0,9 mmHg) Hustota: 1,237 Bod vzplanutí: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070795 CAS číslo: 105728-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...