(±)-γ-Valerolactone ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
(±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone
A18872.14
A18872.22
A18872.36
A18872.14EA
927
CZK
A18872.14
A18872.22
A18872.36
(±)-γ-Valerolactone ≥98%
(±)-γ-Valerolactone
Purity: 98+%
CAS No.: 108-29-2
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 81°(178°F)
Refractive Index: 1.4330
Odour: Pleasant
Melting Point: -31°
Boiling Point: 207-208°
Beilstein Reference: 80420
MDL No.: MFCD00005400
Vzorec:
C₅H₈O₂ MW: 100.12 Bod varu: 207…208 °C Bod tání: –31 °C Hustota: 1.052 Bod vzplanutí: 81 °C (178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005400 CAS číslo: 108-29-2 EINECS: 203-569-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...