2-Fluoro-4-(trifluoromethyl)benzaldehyde ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A18500.03
A18500.06
A18500.14
A18500.03EA
1190
CZK
A18500.03
A18500.06
A18500.14
2-Fluoro-4-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde
Purity: 97%
CAS No.: 89763-93-9
Molecular Formula: C8H4F4O
Formula Weight: 192.11
Flash Point: 110° (230°F)
Refractive Index: 1.4500
Boiling Point: 118-119°
Beilstein Reference: 7703632
MDL No.: MFCD00061310
Vzorec:
C₈H₄F₄O Bod varu: 118…119 °C Hustota: 1,410 Bod vzplanutí: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061310 CAS číslo: 89763-93-9 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...