Tisk…

(Iodomethyl)trimethylammonium iodide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Iodomethylenetrimethylammonium iodide

A18424.06 A18424.14
A18424.06EA 1180 CZK
A18424.06 A18424.14
(Iodomethyl)trimethylammonium iodide ≥98%
(Iodomethyl)trimethylammonium iodide

Purity: 98+% CAS No.: 39741-91-8 Molecular Formula: (CH3)3N(CH2I)I Formula Weight: 326.94 Melting Point: ca 190° dec. Beilstein Reference: 3912137

Vzorec: C₄H₁₁I₂N
Storage Temperature: Ambient
MDL Number: MFCD00031647
CAS číslo: 39741-91-8
EINECS: 254-611-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR