Tisk…

meso-Tetraphenylporphine low chlorine

Dodavatel: Thermo Fisher Scientific

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP

A18261.06 A18261.03 A18261.14
A18261.06EA 7970 CZK
A18261.06 A18261.03 A18261.14
meso-Tetraphenylporphine low chlorine
meso-Tetraphenylporphine

CAS No.: 917-23-7 Molecular Formula: C44H30N4 Formula Weight: 614.71 Melting Point: 449-451° Beilstein Reference: 379542 MDL No.: MFCD00011680

Vzorec: C₄₄H₃₀N₄
Bod tání: 449…451 °C
Hustota: 1,276 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011680
CAS číslo: 917-23-7
EINECS: 213-025-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR