Tisk…

Dithizon ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A17947.14 A17947.18 A17947.30
A17947.14EA 3330 CZK
A17947.14 A17947.18 A17947.30
Dithizon ≥98%
Dithizon

Purity: 98% CAS No.: 60-10-6 Molecular Formula: C13H12N4S Formula Weight: 256.33 Odour: Odourless Melting Point: ca 168° dec. Beilstein Reference: 748838 Merck Reference: 14,3377 MDL No.: MFCD00003025

Vzorec: C₆H₅NHNHCSN=NC₆H₅
Bod varu: 406 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003025
CAS číslo: 60-10-6
EINECS: 200-454-1
Merck Index: 14,3377

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR