Tisk…

1,1,1-Trifluoroiodoethane ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Iodo-2,2,2-trifluoroethane, 2,2,2-Trifluoroethyliodide, 2-Iodo-1,1,1-trifluoroethane, 2,2,2-Trifluoroiodoethane, 1,1,1-Trifluoro-2-iodoethane

A17804.09 A17804.18
A17804.09EA 2080 CZK
A17804.09 A17804.18
1,1,1-Trifluoroiodoethane ≥99%
1,1,1-Trifluoroiodoethane

Purity: 99% CAS No.: 353-83-3 Molecular Formula: CF3CH2I Formula Weight: 209.94 Refractive Index: 1.4009 Boiling Point: 53-55° MDL No.: MFCD00001082

Vzorec: C₂H₂F₃I
MW: 209.94
Bod varu: 53…55 °C
Hustota: 2.142
Storage Temperature: Ambient
MDL Number: MFCD00001082
CAS číslo: 353-83-3
EINECS: 206-541-0
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR