2-Chlorobenzothiazole ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A17762.06
A17762.14
A17762.22
A17762.06EA
1180
CZK
A17762.06
A17762.14
A17762.22
2-Chlorobenzothiazole ≥98%
2-Chlorbenzothiazol
Purity: 98+%
CAS No.: 615-20-3
Molecular Formula: C7H4ClNS
Formula Weight: 169.63
Flash Point:
Vzorec:
C₇H₄ClNS MW: 169,63 g/mol Bod varu: 141 °C (30 mmHg) Bod tání: 30…33 °C Hustota: 1,370 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005776 CAS číslo: 615-20-3 EINECS: 210-415-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...