m-Tolylacetic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(m-Tolyl)acetic acid, (3-Methylphenyl)acetic acid
A17743.09
A17743.18
A17743.30
A17743.09EA
1160
CZK
A17743.09
A17743.18
A17743.30
m-Tolylacetic acid ≥99%
m-Tolylacetic acid
Purity: 99%
CAS No.: 621-36-3
Molecular Formula: C9H10O2
Formula Weight: 150.18
Melting Point: 62-66°
Beilstein Reference: 2043549
Vzorec:
(C₆H₄(CH₃))CH₂COOH MW: 150,18 g/mol Bod varu: 120…123 °C (26 mmHg) Bod tání: 62…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004340 CAS číslo: 621-36-3 EINECS: 210-683-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...