Tisk…

1,3-Bis(diethylamino)propane 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N1,N1,N3,N3-Tetraethylpropane-1,3-diamine, N,N,N',N'-Tetraethyl-1,3-diaminopropane, N,N,N',N'-Tetraethyltrimethylenediamine, N,N,N',N'-Tetraethyl-1,3-propanediamine

A17680.06 A17680.14
A17680.06EA 1730 CZK
A17680.06 A17680.14
1,3-Bis(diethylamino)propane 97%
1,3-Bis(diethylamino)propane

Purity: 97% CAS No.: 60558-96-5 Molecular Formula: (CH3CH2)2N(CH2)3N(CH2CH3)2 Formula Weight: 186.34 Flash Point: 76°(168°F) Refractive Index: 1.4377 Boiling Point: 133°/80mm Beilstein Reference: 1741257 UN No.: 2735 MDL No.: MFCD00009057

Vzorec: C₁₁H₂₆N₂
MW: 186,34 g/mol
Bod varu: 133 °C (80 mmHg)
Hustota: 0,810
Bod vzplanutí: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009057
CAS číslo: 60558-96-5
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR