Tisk…

3-Phenoxyaniline ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Amino-3-phenoxybenzol

A17495.06 A17495.14 A17495.22
A17495.06EA 1530 CZK
A17495.06 A17495.14 A17495.22
3-Phenoxyaniline ≥98%
3-Phenoxyaniline

Purity: 98% CAS No.: 3586-12-7 Molecular Formula: C12H11NO Formula Weight: 185.23 Flash Point: 110°(230°F) Refractive Index: 1.1583 Melting Point: 35-37° Boiling Point: 329-330° MDL No.: MFCD00041891

Vzorec: C₁₂H₁₁NO
MW: 185,23 g/mol
Bod varu: 329…330 °C
Bod tání: 35…37 °C
Hustota: 1,1583
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041891
CAS číslo: 3586-12-7
EINECS: 222-715-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR