Tisk…

Tetrabutylammonium tetrahydroborate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate

A17494.09 A17494.18 A17494.30
A17494.09EA 2680 CZK
A17494.09 A17494.18 A17494.30
Tetrabutylammonium tetrahydroborate ≥98%
Tetrabutylammonium tetrahydroborate

Purity: 98% CAS No.: 33725-74-5 Molecular Formula: C16H40BN Formula Weight: 257.31 Flash Point: 140°(284°F) Melting Point: 125-131° UN No.: 3131 MDL No.: MFCD00012035

Vzorec: C₁₆H₄₀BN
MW: 257,31 g/mol
Bod tání: 125…131 °C
Bod vzplanutí: 140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012035
CAS číslo: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR