1,1,4,4-Tetraphenyl-1,3-butadiene ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
A17225.06
A17225.03
A17225.06EA
4530
CZK
A17225.06
A17225.03
1,1,4,4-Tetraphenyl-1,3-butadiene ≥99%
1,1,4,4-Tetraphenyl-1,3-butadiene
Purity: 99%
CAS No.: 1450-63-1
Molecular Formula: C28H22
Formula Weight: 358.48
Melting Point: 197-202°
MDL No.: MFCD00004766
Vzorec:
(C₆H₅)₂CCHCHC(C₆H₅)₂ MW: 358,48 g/mol Bod varu: 556 °C (1013 hPa) Bod tání: 197…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004766 CAS číslo: 1450-63-1 EINECS: 215-914-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...