4-Aminophthalimide 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Aminoisoindoline-1,3-dione, 5-Amino-isoindole-1,3-dione
A17051.06
A17051.14
A17051.06EA
2580
CZK
A17051.06
A17051.14
4-Aminophthalimide 97%
4-Aminophthalimide
Purity: 97%
CAS No.: 3676-85-5
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Melting Point: 289-291°
MDL No.: MFCD00041854
Vzorec:
C₈H₆N₂O₂ MW: 162,15 g/mol Bod varu: 447 °C (1013 hPa) Bod tání: 289…291 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041854 CAS číslo: 3676-85-5 EINECS: 222-948-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...