α,α,α-Trifluoro-m-toluonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Cyanobenzotrifluoride, m-(Trifluoromethyl)benzonitrile, 3-(Trifluoromethyl)benzonitrile
A17029.06
A17029.14
A17029.22
A17029.06EA
773
CZK
A17029.06
A17029.14
A17029.22
α,α,α-Trifluoro-m-toluonitrile ≥98%
α,α,α-Trifluoro-m-toluonitrile
Purity: 98%
CAS No.: 368-77-4
Molecular Formula: C8H4F3N
Formula Weight: 171.12
Flash Point: 72°(161°F)
Refractive Index: 1.4590
Melting Point: 14-18°
Boiling Point: 188-190°
Beilstein Reference: 1868102
UN No.: 3276
MDL No.: MFCD00001807
Vzorec:
C₈H₄F₃N MW: 171,12 g/mol Bod varu: 188…190 °C Bod tání: 14…18 °C Hustota: 1,280 Bod vzplanutí: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001807 CAS číslo: 368-77-4 EINECS: 206-711-4 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...