Tisk…

Sodium trichloroacetate ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A17004.09 A17004.0B A17004.0I A17004.30
A17004.09EA 749 CZK
A17004.09 A17004.0B A17004.0I A17004.30
Sodium trichloroacetate ≥97%
Sodium trichloroacetate

Purity: 97% CAS No.: 650-51-1 Molecular Formula: CCl3CO2Na Formula Weight: 185.37 Melting Point: 300° Beilstein Reference: 3572664 Merck Reference: 14,9627 UN No.: 3077 MDL No.: MFCD00064198

Vzorec: C₂Cl₃NaO₂
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064198
CAS číslo: 650-51-1
EINECS: 211-479-2
UN: 3077
ADR: 9,III
Merck Index: 14,9627

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR