3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A16880.09
A16880.18
A16880.09EA
1140
CZK
A16880.09
A16880.18
3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98%
3,5-Di-tert-butyl-4-hydroxybenzoic acid
Purity: 98%
CAS No.: 1421-49-4
Molecular Formula: C15H22O3
Formula Weight: 250.34
Melting Point: 206-209°
Beilstein Reference: 2216948
Vzorec:
C₁₅H₂₂O₃ MW: 250,34 g/mol Bod varu: 374 °C (1013 hPa) Bod tání: 206…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008827 CAS číslo: 1421-49-4 EINECS: 215-823-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...