1,3-Dibutyl-2-thiourea ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N,N'-Dibutylthiourea, 1,3-Dibutylthiourea, N,N'-Di-n-butylthiourea
A16783.0B
A16783.18
A16783.30
A16783.0BEA
5360
CZK
A16783.0B
A16783.18
A16783.30
1,3-Dibutyl-2-thiourea ≥98%
1,3-Dibutyl-2-thiourea
Purity: 98%
CAS No.: 109-46-6
Molecular Formula: CH3(CH2)3NHCSNH(CH2)3CH3
Formula Weight: 188.34
Odour: Odourless
Melting Point: 64-67°
Beilstein Reference: 507434
MDL No.: MFCD00004926
Vzorec:
H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃ MW: 188,34 g/mol Bod varu: 252 °C (1013 hPa) Bod tání: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004926 CAS číslo: 109-46-6 EINECS: 203-674-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...