3-Benzoylpropionic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid
A16636.14
A16636.22
A16636.14EA
1530
CZK
A16636.14
A16636.22
3-Benzoylpropionic acid ≥98%
3-Benzoylpropionic acid
Purity: 98+%
CAS No.: 2051-95-8
Molecular Formula: C6H5COCH2CH2CO2H
Formula Weight: 178.19
Melting Point: 114-120°
Beilstein Reference: 639757
Vzorec:
C₁₀H₁₀O₃ MW: 178,19 g/mol Bod varu: 325 °C (1013 hPa) Bod tání: 114…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002792 CAS číslo: 2051-95-8 EINECS: 218-135-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...