Tisk…

Octyl-4-hydroxybenzoate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Octyl parahydroxybenzoate, Octylparaben, 4-Hydroxybenzoic acid octyl ester, n-Octyl 4-hydroxybenzoate

A15964.06 A15964.14 A15964.22
A15964.06EA 989 CZK
A15964.06 A15964.14 A15964.22
Octyl-4-hydroxybenzoate ≥98%
Octyl-4-hydroxybenzoate

Purity: 98% CAS No.: 1219-38-1 Molecular Formula: C15H22O3 Formula Weight: 250.34 Melting Point: 50-53° MDL No.: MFCD00016482

Vzorec: C₁₅H₂₂O₃
Bod tání: 50…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00016482
CAS číslo: 1219-38-1
EINECS: 214-943-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR