Tisk…

o-Tolylacetic acid ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Tolylacetic acid, (2-Methylphenyl)acetic acid

A15783.06 A15783.14 A15783.22
A15783.06EA 910 CZK
A15783.06 A15783.14 A15783.22
o-Tolylacetic acid ≥99%
o-Tolylacetic acid

Purity: 99% CAS No.: 644-36-0 Molecular Formula: C9H10O2 Formula Weight: 150.18 Melting Point: 86-90° Beilstein Reference: 1936952

Vzorec: (C₆H₄CH₃)CH₂COOH
MW: 150,18 g/mol
Bod varu: 283 °C (1013 hPa)
Bod tání: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004328
CAS číslo: 644-36-0
EINECS: 211-416-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR