Tisk…

2-Thiophenemethanol ≥97%

Dodavatel: Thermo Scientific

Synonyms: Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol

A15768.14 A15768.22
A15768.14EA 1520 CZK
A15768.14 A15768.22
2-Thiophenemethanol ≥97%
2-Thiophenemethanol

Purity: 97% CAS No.: 636-72-6 Molecular Formula: C5H6OS Formula Weight: 114.17 Flash Point: 110°(230°F) Refractive Index: 1.5650 Melting Point: 0° Boiling Point: 206-207° Beilstein Reference: 383595 MDL No.: MFCD00005454

Vzorec: C₅H₆OS
MW: 114,17 g/mol
Bod varu: 206…207 °C
Bod tání: 0 °C
Hustota: 1,206
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005454
CAS číslo: 636-72-6
EINECS: 211-264-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR