Tisk…

3-Thenoic acid ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Thiophenecarboxylic acid, Thiophene-3-carboxylic acid

A15712.14 A15712.06 A15712.22
A15712.14EA 9170 CZK
A15712.14 A15712.06 A15712.22
3-Thenoic acid ≥99%
3-Thenoic acid

Purity: 99% CAS No.: 88-13-1 Molecular Formula: C5H4O2S Formula Weight: 128.15 Melting Point: 137-141° Beilstein Reference: 1994 Merck Reference: 14,9279 MDL No.: MFCD00005467

Vzorec: C₅H₄O₂S
MW: 128,15 g/mol
Bod tání: 137…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00005467
CAS číslo: 88-13-1
EINECS: 201-802-5
Merck Index: 14,9279

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR