Tisk…

Methyl-o-toluate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methyl 2-methylbenzoate, o-Toluic acid methyl ester, 2-Toluic acid methyl ester, Methyl-2-methylbenzoate, 2-Methylbenzoic acid methyl ester

A15695.14 A15695.22 A15695.36
A15695.14EA 1140 CZK
A15695.14 A15695.22 A15695.36
Methyl-o-toluate ≥99%
Methyl-o-toluate

Purity: 99% CAS No.: 89-71-4 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 82°(179°F) Refractive Index: 1.5190 Boiling Point: 207-208° Beilstein Reference: 2041846 MDL No.: MFCD00008428

Vzorec: C₉H₁₀O₂
Bod varu: 207…208 °C
Bod tání: –50 °C
Hustota: 1,072
Bod vzplanutí: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008428
CAS číslo: 89-71-4
EINECS: 201-932-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR