Tisk…

4-Chloro-m-phenylenediamine 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Chloro-2,4-diaminobenzene, 4-Chlorobenzene-1,3-diamine, 4-Chloro-1,3-diaminobenzene, 4-Chloro-1,3-phenylenediamine

A15430.06 A15430.14
A15430.06EA 2240 CZK
A15430.06 A15430.14
4-Chloro-m-phenylenediamine 97%
4-Chloro-m-phenylenediamine

Purity: 97% CAS No.: 5131-60-2 Molecular Formula: C6H7ClN2 Formula Weight: 142.59 Melting Point: 87-90° MDL No.: MFCD00025284

Vzorec: C₆H₇ClN₂
MW: 142,59 g/mol
Bod varu: 280 °C (1013 hPa)
Bod tání: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00025284
CAS číslo: 5131-60-2
EINECS: 225-877-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR