3-Nitrophthalimide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A15291.06
A15291.14
A15291.22
A15291.06EA
1260
CZK
A15291.06
A15291.14
A15291.22
3-Nitrophthalimide ≥98%
3-Nitrophthalimide
Purity: 98%
CAS No.: 603-62-3
Molecular Formula: C8H4N2O4
Formula Weight: 192.13
Melting Point: 217-221°
Beilstein Reference: 179965
Vzorec:
C₈H₄N₂O₄ MW: 192,13 g/mol Bod tání: 217…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041852 CAS číslo: 603-62-3 EINECS: 210-051-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...