Tisk…

Furfuryl acetate ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Furan-2-ylmethyl acetate, Acetic acid furfuryl ester

A15285.14 A15285.22
A15285.14EA 948 CZK
A15285.14 A15285.22
Furfuryl acetate ≥97%
Furfuryl acetate

Purity: 97% CAS No.: 623-17-6 Molecular Formula: C7H8O3 Formula Weight: 140.14 Flash Point: 65°(149°F) Refractive Index: 1.4630 Boiling Point: 175-177° Beilstein Reference: 116128 MDL No.: MFCD00003251

Vzorec: C₇H₈O₃
MW: 140.14
Bod varu: 175…177 °C
Hustota: 1.118
Bod vzplanutí: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003251
CAS číslo: 623-17-6
EINECS: 210-775-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR