4-Formylbenzoic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Terephthalaldehydic acid, 4-Carboxybenzaldehyde
A15277.09
A15277.18
A15277.30
A15277.09EA
1400
CZK
A15277.09
A15277.18
A15277.30
4-Formylbenzoic acid ≥98%
4-Formylbenzoic acid
Purity: 98%
CAS No.: 619-66-9
Molecular Formula: C8H6O3
Formula Weight: 150.13
Odour: Odourless
Melting Point: ca 245°
Beilstein Reference: 471734
MDL No.: MFCD00006951
Vzorec:
C₈H₆O₃ MW: 150,13 g/mol Bod varu: 306 °C (1013 hPa) Bod tání: ∼ 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006951 CAS číslo: 619-66-9 EINECS: 210-607-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...