p-Tolylacetic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A15129.06
A15129.14
A15129.22
A15129.06EA
803
CZK
A15129.06
A15129.14
A15129.22
p-Tolylacetic acid ≥99%
p-Tolylacetic acid
Purity: 99%
CAS No.: 622-47-9
Molecular Formula: C9H10O2
Formula Weight: 150.18
Melting Point: 90-93°
Boiling Point: 265-267°
MDL No.: MFCD00004353
Vzorec:
(C₆H₄(CH₃))CH₂COOH Bod varu: 265…267 °C Bod tání: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004353 CAS číslo: 622-47-9 EINECS: 210-738-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...