Tisk…

(4-Methoxyphenyl)acetonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15056.14 A15056.22
A15056.14EA 858 CZK
A15056.14 A15056.22
(4-Methoxyphenyl)acetonitrile ≥98%
(4-Methoxyphenyl)acetonitrile

Purity: 98% CAS No.: 104-47-2 Molecular Formula: C9H9NO Formula Weight: 147.18 Flash Point: 117°(242°F) Refractive Index: 1.5300 Melting Point: 8° Boiling Point: 285-287° Beilstein Reference: 509162 UN No.: 3276 MDL No.: MFCD00001919

Vzorec: C₉H₉NO
Bod varu: 285…287 °C
Bod tání: 8 °C
Hustota: 1,085
Bod vzplanutí: 117 °C (242 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001919
CAS číslo: 104-47-2
EINECS: 203-206-0
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR