α,α,α-Trifluoro-o-toluonitrile ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 97%
CAS No.: 447-60-9
Molecular Formula: C8H4F3N
Formula Weight: 171.12
Flash Point: 90°(194°F)
Refractive Index: 1.4632
Melting Point: 17-18°
Boiling Point: 204-206°
Beilstein Reference: 2048152
UN No.: 3276
MDL No.: MFCD00001791
Vzorec:
C₈H₄F₃N Bod varu: 204…206 °C Bod tání: 17…18 °C Hustota: 1,294 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001791 CAS číslo: 447-60-9 EINECS: 207-184-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...