Tisk…

tert-Butyl bromoacetate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Bromoacetic tert-butyl ester

A14917.30 A14917.18 A14917.09
A14917.30EA 11950 CZK
A14917.30 A14917.18 A14917.09
tert-Butyl bromoacetate ≥98%
tert-Butyl bromoacetate

Purity: 98% CAS No.: 5292-43-3 Molecular Formula: C6H11BrO2 Formula Weight: 195.06 Flash Point: 49°(120°F) Refractive Index: 1.4450 Odour: Pungent Boiling Point: 175° Beilstein Reference: 1753010 UN No.: 2924 MDL No.: MFCD00000188

Vzorec: BrCH₂COOC(CH₃)₂
MW: 195.06
Bod varu: 175 °C
Hustota: 1.330
Bod vzplanutí: 49 °C (120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000188
CAS číslo: 5292-43-3
EINECS: 226-133-6
UN: 2924
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR