Tisk…

Formylhydrazine ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide

A14901.14 A14901.22
A14901.14EA 2450 CZK
A14901.14 A14901.22
Formylhydrazine ≥98%
Formylhydrazine

Purity: 98% CAS No.: 624-84-0 Molecular Formula: HCONHNH2 Formula Weight: 60.06 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 55-59° Beilstein Reference: 635759 MDL No.: MFCD00007589

Vzorec: CH₄N₂O
MW: 60.06
Bod varu: 90 °C (0.7 mmHg)
Bod tání: 55…59 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007589
CAS číslo: 624-84-0
EINECS: 210-867-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR