2-(4-Hydroxyphenyl)ethanol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
A14891.03
A14891.06
A14891.14
A14891.03EA
1080
CZK
A14891.03
A14891.06
A14891.14
2-(4-Hydroxyphenyl)ethanol ≥98%
2-(4-Hydroxyphenyl)ethanol
Purity: 98%
CAS No.: 501-94-0
Molecular Formula: C8H10O2
Formula Weight: 138.17
Melting Point: 89-92°
Boiling Point: 139°/1mm
MDL No.: MFCD00002902
Vzorec:
C₈H₁₀O₂ MW: 138,17 g/mol Bod varu: 139 °C (1 mmHg) Bod tání: 89…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002902 CAS číslo: 501-94-0 EINECS: 207-930-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...