3-(4-Hydroxyphenyl)propionic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A14567.09
A14567.18
A14567.30
A14567.09EA
1170
CZK
A14567.09
A14567.18
A14567.30
3-(4-Hydroxyphenyl)propionic acid ≥99%
3-(4-Hydroxyphenyl)propionic acid
Purity: 99%
CAS No.: 501-97-3
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 126-131°
Beilstein Reference: 2209841
Vzorec:
C₉H₁₀O₃ Bod varu: 208…210 °C (14 mmHg) Bod tání: 126…131 °C Hustota: 1,336 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002778 CAS číslo: 501-97-3 EINECS: 207-931-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...